Order Entry
United States
ContactUsLinkComponent
Human Recombinant LAP (TGF-beta 1) (from HEK293)
Human Recombinant LAP (TGF-beta 1) (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Human Recombinant LAP (TGF-beta 1) (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4 . Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human LAP (TGF-beta 1), His Tag (Cat. No. LAP-H5245) at 1 μg/mL (100 μL/well) can bind Biotinylated Human ITGAV&ITGB6 Heterodimer Protein, His,Avitag&Tag Free (Cat. No. IT6-H82E4) with a linear range of 2-31 ng/mL (QC tested).
  • Gene ID:
    P01137-1 (C33S)
  • Carrier-Free:
    Y
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    CED|TGF-beta-1|LAP (TGF-beta 1)|LAP (TGFB1)|TGFB|DPD1|LAP|TGFB1
  • Protein/Peptide Name:
    LAP (TGF-beta 1)
  • Purity:
    >95%
  • Molecular Weight:
    40 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4 . Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping Temperature:
    Powder,RT

Specifications

Frequently Bought Together