Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant Integrin alpha V beta 8 (from HEK293)
 
Human Recombinant Integrin alpha V beta 8 (from HEK293)

 

  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH8.0. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human TGFB1, His Tag (Cat. No. TG1-H524x) at 5 μg/mL (100 μL/well) can bind Biotinylated Human ITGAV&ITGB8 Heterodimer Protein, His,Avitag&Tag Free (Cat. No. IT8-H82W5) with a linear range of 5-156 ng/mL (QC tested).
  • Gene ID:
    P06756-1(V783I) (ITGAV) and P26012-1 (ITGB8)
  • Carrier-Free:
    Y
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    ITGAV&ITGB8|Integrin alpha V beta 8
  • Protein/Peptide Name:
    Integrin alpha V beta 8
  • Purity:
    >90%
  • Molecular Weight:
    114.7 kDa (ITGAV) and 76.5 kDa (ITGB8)
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH8.0. Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping Temperature:
    Powder,RT

 

Frequently Bought Together