Order Entry
United States
ContactUsLinkComponent
Human Recombinant Integrin alpha V beta 8 (from HEK293)
Human Recombinant Integrin alpha V beta 8 (from HEK293)
Supplier:  ACROBIOSYSTEMS
Certificates
Human Recombinant Integrin alpha V beta 8 (from HEK293)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM NaCl, pH8.0. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human ITGAV&ITGB8 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT8-H52W4) at 5 μg/mL (100 μL/well) can bind Biotinylated Human Latent TGF-Beta 1, His,Avitag (Cat. No. TG1-H82Qb) with a linear range of 1-63 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Human Fibronectin at 10 μg/mL (100 μL/well) can bind Human ITGAV&ITGB8 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT8-H52W4) with a linear range of 0.313-10 μg/mL (Routinely tested).; Measured by its binding ability in a functional ELISA. Immobilized Human Vitronectin at 5 μg/mL (100 μL/well) can bind Human ITGAV&ITGB8 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT8-H52W4) with a linear range of 0.313-10 μg/mL (Routinely tested).
  • Gene ID:
    P06756-1(V783I) | P26012-1
  • Carrier-Free:
    Y
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    ITGAV&ITGB8|Integrin alpha V beta 8
  • Protein/Peptide Name:
    Integrin alpha V beta 8
  • Purity:
    >95%
  • Molecular Weight:
    112.9 kDa (ITGAV) and 76.5 kDa (ITGB8)
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM NaCl, pH8.0. Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping Temperature:
    Powder,RT

Specifications

Frequently Bought Together