Human Recombinant Integrin alpha 8 beta 1 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Fibronectin at 5 μg/mL (100 μL/well) can bind Biotinylated Human ITGA8&ITGB1 Heterodimer Protein, His,Avitag&Tag Free (Cat. No. IT1-H82Wb) with a linear range of 0.156-5 μg/mL (QC tested).
- Gene ID:P53708-1 (ITGA8) and P05556-1 (ITGB1)
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:Integrin alpha 8 beta 1|ITGA8&ITGB1
- Protein/Peptide Name:Integrin alpha 8 beta 1
- Purity:>90%
- Molecular Weight:116.4 kDa (ITGA8) and 83.7 kDa (ITGB1)
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together