Human Recombinant Integrin alpha 4 beta 7 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Natalizumab at 1 μg/mL (100 μL/well) can bind Human ITGA4&ITGB7 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT7-H52W4) with a linear range of 4-63 ng/mL (QC tested).
- Gene ID:P13612-1 (ITGA4) and P26010-1 (ITGB7)
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:ITGA4&ITGB7|Integrin alpha 4 beta 7
- Protein/Peptide Name:Integrin alpha 4 beta 7
- Purity:>90%
- Molecular Weight:111.4 kDa (ITGA4) and 81.8 kDa (ITGB7)
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together