Human Recombinant IL-7 R alpha (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized ActiveMax® Human IL-7, Tag Free (Cat. No. IL7-H4219) at 2 μg/mL (100 μL/well) can bind Human IL-7 R alpha, Mouse IgG2a Fc Tag (Cat. No. IL7-H5258) with a linear range of 4-31 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Biotinylated Human TSLP (R127A, R130A), His,Avitag (Cat. No. TSP-H82E0) immobilized at 2 μg/mL (100 μL/well) via precoated 5 μg/mL (100 μL/well) of Human TSLP R, Fc Tag (Cat. No. TSR-H525a), can bind can bind Human IL-7 R alpha, Mouse IgG2a Fc Tag (Cat. No. IL7-H5258) with a linear range of 1-20 ng/mL (Routinely tested).; Captured Human IL-7 R alpha, Mouse IgG2a Fc Tag (Cat. No. IL7-H5258) on CM5 chip via anti-mouse antibodies surface can bind ActiveMax® Human IL-7, Tag Free (Cat. No. IL7-H4219) with an affinity constant of 13.3 nM as determined in a SPR assay (Biacore T200) (Routinely tested).
- Gene ID:P16871-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:IL7Ra|CD127
- Protein/Peptide Name:IL-7 R alpha
- Purity:>90%
- Molecular Weight:52.0 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together