Human Recombinant IL-6 R alpha (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human IL-6, Tag Free (Cat. No. IL6-H4218) at 5 μg/mL (100 μL/well) can bind Biotinylated Human IL-6 R alpha, Avitag,His Tag (Cat. No. CD6-H82E8) with a linear range of 4-125 ng/mL (QC tested).
- Gene ID:P08887-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:IL-6RA|IL6RA|IL-6R-alpha|IL6RQ|IL-6R-1|IL6R|CD126|gp80
- Protein/Peptide Name:IL-6 R alpha
- Purity:>95%
- Molecular Weight:41.7 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together