Human Recombinant IL-4 R alpha (from HEK293)
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human IL-4, Tag Free (Cat. No. IL4-H4218) at 5 μg/mL (100 μL/well) can bind Biotinylated Human IL-4 R alpha, Avitag,His Tag (Cat. No. ILR-H82E9) with a linear range of 0.005-0.078 μg/mL (QC tested).
- Gene ID:P24394
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:IL4R|IL4RA|CD124
- Protein/Peptide Name:IL-4 R alpha
- Purity:>95%
- Molecular Weight:26.9 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Frequently Bought Together