Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant IL-3 R alpha (from HEK293)
 
Human Recombinant IL-3 R alpha (from HEK293)

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Immobilized Human IL-3 R alpha, His Tag (SPR verified) (Cat. No. ILA-H52H6) on CM5 Chip via anti-His antibody can bind Human IL-3, Tag Free with an affinity constant of 224 nM as determined in SPR assay (Biacore T200) (QC tested).; Loaded Human IL-3 R alpha, His Tag (SPR verified) (Cat. No. ILA-H52H6) on NTA Biosensor, can bind Human IL-3, Tag Free with an affinity constant of 370 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    P26951-1
  • Carrier-Free:
    Y
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    IL3RX|IL3RA|CD123 antigen|IL-3Ra|IL3RY|CD123|IL-3 R alpha|hIL3Ra|MGC34174|hIL-3Ra|IL3RAY|IL3R|IL-3R-alpha
  • Protein/Peptide Name:
    IL-3 R alpha
  • Purity:
    >95%
  • Molecular Weight:
    35.0 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping Temperature:
    Powder,RT

 

Frequently Bought Together