Human Recombinant IL-17A and IL-17F (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Anti-Human IL17A MAb at 2 μg/mL (100 μL/well) can bind Biotinylated Human IL-17A&IL-17F, Avitag,His Tag (Cat. No. ILF-H82W1) with a linear range of 0.2-6 ng/mL (QC tested).
- Gene ID:Q16552-1 (IL-17A) and Q96PD4-1 (IL-17F)
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:IL-17|IL24|IL-17F|IL-17A|ML-1|Interleukin-17F|CTLA-8|Interleukin-17A
- Protein/Peptide Name:IL-17A and IL-17F
- Purity:>95%
- Molecular Weight:34.1 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together