Human Recombinant IL-12B and IL-12A (from HEK293)
Catalog # 103739-242
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:20 µg
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human IL-12B&IL-12A Heterodimer Protein, His Tag&Flag Tag (Cat. No. IL2-H4210) at 0.5 μg/mL (100 μL/well) can bind Anti-IL23A and IL12B MAb (P40 domain) with a linear range of 0.2-2 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Biotinylated Human IL-12 R beta 1, Fc,Avitag (Cat. No. ILB-H82F7) at 1 μg/mL (100 μL/well) on Streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate, can bind Human IL-12B&IL-12A Heterodimer Protein, His Tag&Flag Tag (Cat. No. IL2-H4210) with a linear range of 0.039-0.625 μg/mL (Routinely tested).; Human IL-12 R beta 1, Fc Tag (Cat. No. ILB-H5255) captured on CM5 chip via Anti-human IgG Fc antibodies surface can bind Human IL-12B&IL-12A Heterodimer Protein, His Tag&Flag Tag (Cat. No. IL2-H4210) with an affinity constant of 2.06 nM as determined in a SPR assay (Biacore T200) (Routinely tested).; Loaded Anti-IL23A and IL12B MAb (P40 domain) on Protein A Biosensor, can bind Human IL-12B&IL-12A Heterodimer Protein, His Tag&Flag Tag (Cat. No. IL2-H4210) with an affinity constant of 1.07 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; Loaded Human IL-12B&IL-12A Heterodimer Protein, His Tag&Flag Tag (Cat. No. IL2-H4210) on HIS1K Biosensor, can bind Human IL-12 R beta 1, Fc Tag (Cat. No. ILB-H5255) with an affinity constant of 92.2 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; Human IL-12B&IL-12A Heterodimer Protein, His Tag&Flag Tag (Cat. No. IL2-H4210) stimulates the proliferation of PHA-P activated PBMC. The ED50 for this effect is 0.05 ng/mL (Routinely tested).
- Gene ID:P29460-1 (IL12B) and P29459-1 (IL12A)
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:IL12|p70|Interleukin-12
- Protein/Peptide Name:IL-12B & IL-12A
- Purity:>90%
- Molecular Weight:36.6 kDa (IL12B) and 23.8 kDa (IL12A)
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
- Cat. No.:103739-242