Human Recombinant HVEM (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.1 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human HVEM, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. HVM-H5255) at 5 μg/mL (100 μL/well) can bind Human BTLA, Fc Tag (Cat. No. BTA-H5255) with a linear range of 0.156-1.25 μg/mL (QC tested).
- Gene ID:Q92956-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:HVEA|HVEM|CD270|ATAR|TNFRSF14|TR2|LIGHTR
- Protein/Peptide Name:HVEM
- Purity:>95%
- Molecular Weight:44.4 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together