Human Recombinant GM-CSF (from HEK293)
Catalog # 103737-788
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:20 µg
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.1 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Loaded Human GM-CSF R alpha, Fc Tag (Cat. No. GRA-H5255) on Protein A Biosensor, can bind ActiveMax® Human GM-CSF, Tag Free (Cat. No. GMF-H4214) with an affinity constant of 9.21 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; The bio-activity was determined by dose-dependent stimulation of the proliferation of TF-1 cells. The ED50 < 0.1 ng/mL, corresponding to a specific activity of > 1x107 Unit/mg.
- Gene ID:P04141
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:GM-CSF|MGC131935|CSF2
- Protein/Peptide Name:GM-CSF
- Purity:>95%
- Molecular Weight:14.5 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
- Cat. No.:103737-788