Human Recombinant GITR Ligand (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human GITR, Fc Tag (Cat. No. GIR-H5254) at 2 μg/mL (100 μL/well) can bind Biotinylated Human GITR Ligand, Avitag,Fc Tag (Cat. No. GIL-H82F8) with a linear range of 4-63 ng/mL (QC tested).
- Gene ID:AAH69319.1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:hGITRL|TL6|AITRL|TNFSF18|GITR Ligand
- Protein/Peptide Name:GITR Ligand
- Purity:>90%
- Molecular Weight:43.5 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together