Human Recombinant Fc gamma RI / CD64 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Biotinylated Human CD64, His,Avitag (Cat. No. FCA-H82E8) captured on Biotin CAP - Series S sensor Chip can bind Herceptin® with an affinity constant of 1.66 nM as determined in a SPR assay (Biacore T200) (QC tested).; Loaded Biotinylated Human CD64, His,Avitag (Cat. No. FCA-H82E8) on SA Biosensor, can bind Herceptin with an affinity constant of 0.515 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:P12314-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:FCGR1|FCG1|FCRI|FCGR1A|IGFR1|CD64|CD64A
- Protein/Peptide Name:Fc gamma RI / CD64
- Purity:>95%
- Molecular Weight:34.2 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together