Human Recombinant EGF R (from HEK293)
Catalog # 103737-696
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:100 µg
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Erbitux (Cetuximab) captured on CM5 chip via anti-human IgG Fc antibodies surface, can bind Human EGF R, His Tag (Cat. No. EGR-H5222) with an affinity constant of 1.3 nM as determined in a SPR assay (Biacore T200) (Routinely tested).; Loaded Erbitux (Cetuximab) on AHC Biosensor, can bind Human EGF R, His Tag (Cat. No. EGR-H5222) with an affinity constant of 1.23 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; 2e5 of Anti-EGFR CAR-293 cells were stained with 100 μL of 1 μg/mL of Human EGF R, His Tag (Cat. No. EGR-H5222) and negative control protein respectively, washed and then followed by PE anti-His antibody and analyzed with FACS (Routinely tested).
- Gene ID:P00533-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:PIG61|HER1|mENA|EGFR|ERBB|ERBB1
- Protein/Peptide Name:EGF R
- Purity:>95%
- Molecular Weight:70.5kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
- Cat. No.:103737-696