Human Recombinant CEACAM-5 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 20 mM Tris, 150 mM Potassium glutamate, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Gene ID:P06731-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:Meconium antigen 100|CEA|CD66e|CEACAM-5
- Protein/Peptide Name:CEACAM-5
- Purity:>90%
- Molecular Weight:75.0 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 20 mM Tris, 150 mM Potassium glutamate, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together