Human Recombinant CD47 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 10 mM Bicine, pH8.3. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human SIRP alpha, Fc Tag (Cat. No. SIA-H5251) at 2 μg/mL (100 μL/well) can bind Unconjugated Human CD47, His,Avitag (Cat. No. CD7-HA2E9) with a linear range of 2-13 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Anti-CD47 MAb, Human IgG4 at 2 μg/mL (100 μL/well) can bind Unconjugated Human CD47, His,Avitag (Cat. No. CD7-HA2E9) with a linear range of 0.05-2 ng/mL (QC tested).
- Gene ID:Q08722-3
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:IAP|MER6|OA3
- Protein/Peptide Name:CD47
- Purity:>95%
- Molecular Weight:16.9 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 10 mM Bicine, pH8.3. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together