Human Recombinant CD40 Ligand (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human CD40, His Tag (Cat. No. CD0-H5228) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD40 Ligand, Avitag,Fc Tag (Cat. No. CDL-H82F1) with a linear range of 1-31 ng/mL (QC tested).
- Gene ID:P29965-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:TRAP|CD40L|IMD3|gp39|CD154|CD40LG|IGM|T-BAM|TNFSF5|HIGM1
- Protein/Peptide Name:CD40 Ligand
- Purity:>95%
- Molecular Weight:45.0 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together