Human Recombinant CD30 (from HEK293)
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human CD30 Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CDL-H525b) at 5 μg/mL (100 μL/well) can bind Biotinylated Human CD30, Avitag,His Tag (Cat. No. CD0-H82E6) with a linear range of 10-78 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Anti-CD30 MAb, Human IgG1 at 5 μg/mL (100 μL/well) can bind Biotinylated Human CD30, Avitag,His Tag (Cat. No. CD0-H82E6) with a linear range of 0.4-6.5 ng/mL (Routinely tested).; Loaded Biotinylated Human CD30, Avitag,His Tag (Cat. No. CD0-H82E6) on SA Biosensor, can bind Human CD30 Ligand, His Tag, low endotoxin (Cat. No. CDL-H524b) with an affinity constant of 132 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:P28908
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:Ki-1|CD30|D1S166E|TNFRSF8
- Protein/Peptide Name:CD30
- Purity:>95%
- Molecular Weight:41.1 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Frequently Bought Together