Human Recombinant CD27 Ligand (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human CD27, Fc Tag (Cat. No. CD7-H5254) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD27 Ligand, Avitag,Fc Tag (Cat. No. TN7-H82F4) with a linear range of 1-31 ng/mL (QC tested).
- Gene ID:P32970-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:CD27LG|TNFSF7|TNFSF7G|CD27L|CD70
- Protein/Peptide Name:CD27 Ligand
- Purity:>85%
- Molecular Weight:45 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together