Order Entry
United States
ContactUsLinkComponent
Human Recombinant CD27 (from HEK293)
Human Recombinant CD27 (from HEK293)
Supplier:  ACROBIOSYSTEMS
Certificates
Human Recombinant CD27 (from HEK293)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human CD27 Ligand, Fc Tag (Cat. No. TN7-H526x) at 10 μg/mL (100 μL/well) can bind Biotinylated Human CD27, Fc,Avitag (Cat. No. TN7-H82F6) with a linear range of 5-156 ng/mL (QC tested).; Flow Cytometry assay shows that Biotinylated Human CD27, Fc,Avitag (Cat. No. TN7-H82F6) can bind to Raji cells Surface Human CD70. The concentration of CD27 used is 10 μg/mL (Routinely tested).
  • Gene ID:
    P26842
  • Carrier-Free:
    Y
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    Tp55|S152|T14|TNFRSF7|CD27
  • Protein/Peptide Name:
    CD27
  • Purity:
    >95%
  • Molecular Weight:
    47.7 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping Temperature:
    Powder,RT

Specifications

Frequently Bought Together