Human Recombinant CD27 (from HEK293)
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human CD27, His Tag (Cat. No. CD7-H522b) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD27 Ligand, Avitag,Fc Tag (Cat. No. TN7-H82F4) with a linear range of 2-31 ng/mL (QC tested).
- Gene ID:P26842-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:Tp55|S152|T14|TNFRSF7|CD27
- Protein/Peptide Name:CD27
- Purity:>90%
- Molecular Weight:21.1 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Frequently Bought Together