Human Recombinant CD19 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; FMC63 MAb (mouse lgG2a) captured on CM5 chip via anti-mouse antibodies surface, can bind Human CD19 (20-291), His Tag (Cat. No. CD9-H52H2) with an affinity constant of 2.95 nM as determined in a SPR assay (Biacore T200) (Routinely tested).; Captured Human CD19 (20-291), His Tag (Cat. No. CD9-H52H2) on CM5 Chip via anti-His antibody, can bind FMC63 (mouse lgG2a) with an affinity constant of 2.01 nM as determined in SPR assay (Biacore T200 ) (Routinely tested).; Loaded FMC63 MAb (mouse lgG2a) on Protein A Biosensor, can bind Human CD19 (20-291), His Tag (Cat. No. CD9-H52H2) with an affinity constant of 4.36 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; Loaded Anti-CD19 MAb (Human IgG1) on AHC Biosensor, can bind Human CD19 (20-291), His Tag (Cat. No. CD9-H52H2) with an affinity constant of 5.41 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; 293 cells were transfected with FMC63-scFv and RFP tag. 2e5 of the cells were first incubated with A. Human His Tag control. B. Human CD19 (20-291), His Tag (Cat. No. CD9-H52H2, 10 μg/mL). C. Human CD19 (20-291), His Tag (Cat. No. CD9-H52H2, 10 μg/mL) and FMC63 (Mouse anti-CD19 antibody). The FITC Anti-6xHis tag antibody was used to analyse with FACS. RFP was used to evaluate CAR (FMC63-scFv) expression and FITC was used to evaluate the binding activity of Human CD19 (20-291), His Tag (Cat. No. CD9-H52H2).
- Gene ID:P15391-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:CVID3|MGC12802|B4|CD19
- Protein/Peptide Name:CD19
- Purity:>90%
- Molecular Weight:32.0 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together