Order Entry
United States
ContactUsLinkComponent
Human Recombinant CD155 (from HEK293)
Human Recombinant CD155 (from HEK293)
Supplier:  ACROBIOSYSTEMS
Certificates
Human Recombinant CD155 (from HEK293)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human TIGIT, Fc Tag (Cat. No. TIT-H5254) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD155, Fc,Avitag (Cat. No. CD5-H82F6) with a linear range of 0.5-16 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Human DNAM-1, Fc Tag (Cat. No. DN1-H5257) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD155, Fc,Avitag (Cat. No. CD5-H82F6) with a linear range of 0.02-0.312 μg/mL (Routinely tested).; Serial dilutions of TIGIT antibody antibody (1:2 serial dilutions, from 20 μg/mL to 0.0097 μg/mL) were added into Human TIGIT, His Tag (Cat. No. TIT-H52H3): Biotinylated Human CD155, Fc,Avitag (Cat. No. CD5-H82F6) binding reactions. The assay was performed according to the above described protocol. Background was subtracted from data points before curve fitting.; FACS assay shows that Biotinylated Human CD155, Fc,Avitag (Cat. No. CD5-H82F6) can bind to 293T cell overexpressing human TIGIT. The concentration of CD155 is 1 μg/mL (QC tested).; FACS analysis shows that the binding of Biotinylated Human CD155, Fc,Avitag (Cat. No. CD5-H82F6) to 293T overexpressing TIGIT was inhibited by increasing concentration of neutralizing Anti-Human TIGIT MAb. The concentration of CD155 used is 1 μg/mL. The IC50 is 0.201 μg/mL (Routinely tested).
  • Gene ID:
    P15151 (I340M)
  • Carrier-Free:
    Y
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    TAGE4|PVS|FLJ25946|CD155|NECL5|PVR|HVED
  • Protein/Peptide Name:
    CD155
  • Purity:
    >95%
  • Molecular Weight:
    63.5 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping Temperature:
    Powder,RT

Specifications

Frequently Bought Together