Human Recombinant CD155 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.1 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human TIGIT, Fc Tag (Cat. No. TIT-H5254) at 2 μg/mL (100 μL/well) can bind Human CD155, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD5-H5254) with a linear range of 0.1-4 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Human DNAM-1, Fc Tag (Cat. No. DN1-H5257) at 2 μg/mL (100 μL/well) can bind Human CD155, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD5-H5254) with a linear range of 1-16 ng/mL (Routinely tested).; Serial dilutions of Human TIGIT Neutralizing antibody were added into Human CD155, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD5-H5254): Biotinylated Human TIGIT, Fc,Avitag (Cat. No. TIT-H82F1) binding reactions. The half maximal inhibitory concentration (IC50) is 0.08116 μg/mL (Routinely tested).; Loaded Human CD155, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD5-H5254) on Protein A Biosensor, can bind Human TIGIT, His Tag (Cat. No. TIT-H52H3) with an affinity constant of 0.24 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; Loaded Human CD155, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD5-H5254) on Protein A Biosensor, can bind Human DNAM-1, His Tag (Cat. No. DN1-H52H6) with an affinity constant of 0.57 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; FACS assay shows that Human CD155, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD5-H5254) can bind to 293T cell overexpressing human TIGIT. The concentration of CD155 is 1 μg/mL (Routinely tested).; FACS analysis shows that the binding of Human CD155, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD5-H5254) to 293T overexpressing TIGIT was inhibited by increasing concentration of neutralizing Anti-Human TIGIT MAb. The concentration of CD155 used is 1 μg/mL. The IC50 is 0.1413 μg/mL (Routinely tested).
- Gene ID:P15151-1 (I340M)
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:TAGE4|PVS|FLJ25946|CD155|NECL5|PVR|HVED
- Protein/Peptide Name:CD155
- Purity:>95%
- Molecular Weight:62.2 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together