Human Recombinant CBLB (from E. coli)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 40 mM HEPES, 120 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Gene ID:Q13191-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:RNF56|CBLB|CBL-B
- Protein/Peptide Name:CBLB
- Purity:>90%
- Molecular Weight:47.9 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 40 mM HEPES, 120 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together