Human Recombinant BCMA (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human BAFF, Fc Tag (Cat. No. BAF-H4268) at 5 μg/mL (100 μL/well) can bind Biotinylated Human BCMA, Fc,Avitag (Cat. No. BC7-H82F0) with a linear range of 0.12-1.95 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Anti-Human BCMA Mab at 2 μg/mL (100 μL/well) can bind Biotinylated Human BCMA, Fc,Avitag (Cat. No. BC7-H82F0) with a linear range of 0.1-3 ng/mL (Routinely tested).; 293 cells were transfected with anti-BCMA-scFv and RFP tag. 2e5 of the cells were first stained with B. Biotinylated Human BCMA, Fc,Avitag (Cat. No. BC7-H82F0, 3 μg/ml) and C. biotinylated protein control, followed by FITC-conjugated streptavidin. A. Non-transfected 293 cells and C. biotinylated protein control were used as negative control. RFP was used to evaluate CAR (anti-BCMA-scFv) expression and FITC was used to evaluate the binding activity of Biotinylated Human BCMA, Fc,Avitag (Cat. No. BC7-H82F0).; Human T cells were transfected with anti-BCMA CAR and cultured for 3 days. Three days post-transfection, 1e6 cells were first incubated with 50ul Biotinylated Human BCMA, Fc,Avitag (Cat.No. BC7-H82F0, 8 μg/ml ), washed and then stained with PE Streptavidin and analyzed by flow cytometry. (Data are kindly provided by PREGENE Biopharma)
- Gene ID:Q02223
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:CD269|TNFRSF17|BCM
- Protein/Peptide Name:BCMA
- Purity:>95%
- Molecular Weight:33.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together