Order Entry
United States
ContactUsLinkComponent
Human Recombinant BCMA (from HEK293)
Human Recombinant BCMA (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Human Recombinant BCMA (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Anti-Human BCMA MAb (human IgG1) captured on CM5 chip via Anti-Human IgG Fc antibodies surface, can bind Human BCMA, His Tag (Cat. No. BCA-H522y) with an affinity constant of 32.3 nM as determined in a SPR assay (Biacore T200) (Routinely tested).; Anti-BCMA Neutralizing Antibody, Mouse IgG1 (Clone: AM43, Cat. No. BCA-M43) captured on CM5 chip via anti-mouse antibodies surface can bind Human BCMA, His Tag (Cat. No. BCA-H522y) with an affinity constant of 0.388 nM as determined in a SPR assay (Biacore T200) (Routinely tested).; Loaded Anti-Human BCMA MAb (human IgG1) on Protein A Biosensor, can bind Human BCMA, His Tag (Cat. No. BCA-H522y) with an affinity constant of 35.8 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; Loaded Biotinylated Human BAFF, His,Avitag (active trimer) (MALS verified) (Cat. No. BAF-H82Q2) on SA Biosensor, can bind Human BCMA, His Tag (Cat. No. BCA-H522y) with an affinity constant of 1.1 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; Loaded Biotinylated Human BAFF, Avitag,Fc Tag (active trimer) (MALS verified) (Cat. No. BAF-H82F3) on SA Biosensor, can bind Human BCMA, His Tag (Cat. No. BCA-H522y) with an affinity constant of 1 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; 293 cells were transfected with anti-BCMA-scFv and RFP tag. 2e5 of the cells were first stained with B. Human BCMA, His Tag (Cat. No. BCA-H522y, 0.03 μg/mL) and C. His tag protein control, followed by FITC-conjugated anti-6xHis tag antibody. A. Non-transfected 293 cells and C. His tag protein control were used as negative control. RFP was used to evaluate CAR (anti-BCMA-scFv) expression and FITC was used to evaluate the binding activity of Human BCMA, His Tag (Cat. No. BCA-H522y).
  • Gene ID:
    Q02223-1
  • Carrier-Free:
    Y
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    CD269|TNFRSF17|BCM
  • Protein/Peptide Name:
    BCMA
  • Purity:
    >90%
  • Molecular Weight:
    7.8 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping Temperature:
    Powder,RT

Specifications

Frequently Bought Together