Human Recombinant B7-H7 (from HEK293)
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human CD28H, Fc Tag (Cat. No. CDH-H5251) at 5 μg/mL (100 μL/well) can bind Biotinylated Human B7-H7, Fc,Avitag (Cat. No. B77-H82F5) with a linear range of 5-78 ng/mL (QC tested).
- Gene ID:Q9UM44-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:B7-H7|B7 Homolog 7|HHLA2
- Protein/Peptide Name:B7-H7
- Purity:>90%
- Molecular Weight:65.9 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Frequently Bought Together