Human Recombinant B7-H4 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Anti-Human B7-H4 MAb, Human IgG1 at 2 μg/mL (100 μL/well) can bind Biotinylated Human B7-H4, Avitag,His Tag (recommended for biopanning) (Cat. No. B74-H82E2) with a linear range of 3-48 ng/mL (QC tested).
- Gene ID:Q7Z7D3
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:B7h.5|B7-H4|B7S1|VTCN1
- Protein/Peptide Name:B7-H4
- Purity:>90%
- Molecular Weight:28 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together