Human Recombinant B7-H3 (from HEK293)
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Monoclonal Anti-Human B7-H3 / B7-H3 (4Ig) Antibody, Human IgG1 at 2 μg/mL (100 μL/well) can bind Biotinylated Human B7-H3, Avitag,His Tag (Cat. No. B73-H82E6) with a linear range of 5-39 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Biotinylated Human B7-H3, Avitag,His Tag (Cat. No. B73-H82E6) at 1 μg/mL (100 μL/well) on Streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate, can bind Monoclonal Anti-Human B7-H3 / B7-H3 (4Ig) Antibody, Human IgG1 with a linear range of 0.3-5 ng/mL (Routinely tested).
- Gene ID:Q5ZPR3-2
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:B7 homolog 3|B7-H3|CD276
- Protein/Peptide Name:B7-H3
- Purity:>95%
- Molecular Weight:26.5 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Frequently Bought Together