Human Recombinant B7-1 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Anti-CD80 MAb, Human IgG1 at 5 μg/mL (100 μL/well) can bind Biotinylated Human B7-1, Avitag,His Tag (recommended for biopanning) (Cat. No. B71-H82E9) with a linear range of 0.1-3 ng/mL (QC tested).
- Gene ID:P33681-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:B7-1|B7.1|CD28LG|CD28LG1|LAB7|BB1|CD80|B7
- Protein/Peptide Name:B7-1
- Purity:>95%
- Molecular Weight:26.5 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together