Human Recombinant B7-1 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human CTLA-4, Fc Tag (Cat. No. CT4-H5255) at 5 μg/mL (100 μL/well) can bind Biotinylated Human B7-1, Fc,Avitag (Cat. No. B71-H82F2) with a linear range of 0.6-9.8 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Human PD-L1, Fc Tag (Cat. No. PD1-H5258) at 10 μg/mL (100 μL/well) can bind Biotinylated Human B7-1, Fc,Avitag (Cat. No. B71-H82F2) with a linear range of 0.156-5 μg/mL (Routinely tested).; Serial dilutions of Ipilimumab were added into Human CTLA-4, Fc Tag (Cat. No. CT4-H5255): Biotinylated Human B7-1, Fc,Avitag (Cat. No. B71-H82F2) binding reactions. The half maximal inhibitory concentration (IC50) is 0.8260 μg/mL (Routinely tested).; Flow Cytometry assay shows that Biotinylated Human B7-1, Fc,Avitag (Cat. No. B71-H82F2) can bind to 293 cell overexpressing human CTLA-4. The concentration of Human B7-1 is 0.3 μg/mL (Routinely tested).; FACS analysis shows that the binding of Biotinylated Human B7-1, Fc,Avitag (Cat. No. B71-H82F2) to 293 overexpressing CTLA-4 was inhibited by increasing concentration of neutralizing Anti-Human CTLA-4 antibody. The concentration of B7-1 used is 0.3 μg/mL. The IC50 is 3.025 μg/mL (Routinely tested).
- Gene ID:P33681
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:B7-1|B7.1|CD28LG|CD28LG1|LAB7|BB1|CD80|B7
- Protein/Peptide Name:B7-1
- Purity:>95%
- Molecular Weight:52.3 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together