Human Recombinant APRIL (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human BCMA, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. BCA-H5253) at 5 μg/mL (100 μL/well) can bind Human APRIL, Fc Tag (Cat. No. APL-H5267) with a linear range of 0.6-10 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Human APRIL, Fc Tag (Cat. No. APL-H5267) at 5 μg/mL (100 μL/well) can bind Biotinylated Human TACI, Fc,Avitag (Cat. No. TAI-H82F6) with a linear range of 0.01-0.313 μg/mL (Routinely tested).
- Gene ID:O75888-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:APRIL|TNFSF13|TALL2|TRDL-1|ZTNF2|TALL-2|CD256
- Protein/Peptide Name:APRIL
- Purity:>95%
- Molecular Weight:42.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together