Human Recombinant Activin A (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.1 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 0.056% TFA in 60% ACN. Normally trehalose is added as protectant before lyophilization.; The bio-activity of ActiveMax® Human Activin A, Tag Free, low endotoxin (Cat. No. ACA-H421b) was determined by dose-dependent inhibition of the proliferation of MPC-11 cells. The EC50 for this effect is typically ≤ 9.5 ng/mL (Routinely tested).
- Gene ID:P08476
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:Activin A|INHBA
- Protein/Peptide Name:Activin A
- Purity:>95%
- Molecular Weight:13.0 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 0.056% TFA in 60% ACN. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together