Human Recombinant Activin A (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; The bio-activity of Human Latent Activin A, His Tag (Cat. No. ACA-H424x) was determined by dose-dependent inhibition of the proliferation of MPC-11 cells. The EC50 for this effect is typically 47-66 ng/mL (Routinely tested).
- Gene ID:P08476
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:Activin A|INHBA
- Protein/Peptide Name:Activin A
- Purity:>95%
- Molecular Weight:13.0 kDa (mature) and 32.0 kDa (pro)
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together