Human / Cynomolgus / Rhesus macaque Recombinant CD28 (from HEK293)
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkeyRhesus monkeyHuman
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Biotinylated Human / Cynomolgus / Rhesus macaque CD28, Fc,Avitag (Cat. No. CD8-H82F2) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Human B7-1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. B71-H52A4) with a linear range of 5-78 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Anti-CD28 MAb, Mouse IgG1 at 5 μg/mL (100 μL/well) can bind Biotinylated Human / Cynomolgus / Rhesus macaque CD28, Fc,Avitag (Cat. No. CD8-H82F2) with a linear range of 2.4-39 ng/mL (Routinely tested).; Measured by its binding ability in a functional ELISA. Immobilized Human B7-1, Fc Tag (Cat. No. B71-H5259) at 5 μg/mL (100 μL/well) can bind Biotinylated Human / Cynomolgus / Rhesus macaque CD28, Fc,Avitag (Cat. No. CD8-H82F2) with a linear range of 0.313-5 μg/mL (Routinely tested).
- Gene ID:P10747
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:CD28|Tp44
- Protein/Peptide Name:CD28
- Purity:>95%
- Molecular Weight:43.6 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Frequently Bought Together