Order Entry
United States
ContactUsLinkComponent
HIV-1 (CN54) Recombinant GP120 (from HEK293)
HIV-1 (CN54) Recombinant GP120 (from HEK293)
Supplier:  ACROBIOSYSTEMS
Certificates
HIV-1 (CN54) Recombinant GP120 (from HEK293)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    HIV-1 (CN54)
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human CD4, Fc Tag (Cat. No. CD4-H5259) at 5 μg/mL (100 μL/well) can bind Biotinylated HIV-1 [HIV-1/Clade B/C (CN54)] GP120, His,Avitag (Cat. No. GP0-V182E6) with a linear range of 0.156-1.25 μg/mL (QC tested).
  • Gene ID:
    G4XFJ5-1 (E46G, T396A, A497T)
  • Carrier-Free:
    Y
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    GP120|GP120-CN54
  • Protein/Peptide Name:
    GP120
  • Purity:
    >95%
  • Molecular Weight:
    56.7 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping Temperature:
    Powder,RT

Specifications

Frequently Bought Together