Order Entry
United States
ContactUsLinkComponent
Hamster Recombinant PCSK9 (from HEK293), Acro Biosystems
Hamster Recombinant PCSK9 (from HEK293), Acro Biosystems
Supplier:  ACRO Biosystems
Certificates
Hamster Recombinant PCSK9 (from HEK293), Acro Biosystems
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Hamster
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human LDL R, Strep Tag (Cat. No. LDR-H5281) at 5 μg/mL (100 μL/well) can bind Hamster PCSK9, His Tag (Cat. No. PC9-H52E4) with a linear range of 1-16 ng/mL (QC tested).; Loaded Human LDL R, Fc Tag (Cat. No. LDR-H5254) on Protein A Biosensor, can bind Hamster PCSK9, His Tag (Cat. No. PC9-H52E4) with an affinity constant of 4.09 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    G3GTK5
  • Carrier-Free:
    Y
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    HCHOLA3|PCSK9|LDLCQ1|PC9|NARC1|FH3
  • Protein/Peptide Name:
    PCSK9
  • Purity:
    >95%
  • Molecular Weight:
    13.9 kDa and 59.0 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping Temperature:
    Powder,RT

Specifications

Frequently Bought Together