Cynomolgus Recombinant SIRP alpha (from HEK293)
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Cynomolgus / Rhesus macaque CD47, Fc Tag (Cat. No. CD7-C5252) at 2 μg/mL (100 μL/well) can bind Cynomolgus SIRP alpha, His Tag (Cat. No. SIA-C52H7) with a linear range of 0.313-2.5 μg/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Biotinylated Human CD47, Fc,Avitag (Cat. No. CD7-H82F6) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.2ug/well) plate, can bind Cynomolgus SIRP alpha, His Tag (Cat. No. SIA-C52H7) with a linear range of 0.039-0.625 μg/mL (Routinely tested).
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:MYD1|PTPNS1|CD172A|SHPS1|P84|SIRPA|BIT|MFR
- Protein/Peptide Name:SIRP alpha
- Purity:>95%
- Molecular Weight:39.0 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Frequently Bought Together