Order Entry
United States
ContactUsLinkComponent
Cynomolgus Recombinant FOLR1 (from HEK293)
Cynomolgus Recombinant FOLR1 (from HEK293)
Catalog # 103737-604
Supplier:  ACRO Biosystems
CAS Number:  
Cynomolgus Recombinant FOLR1 (from HEK293)
Catalog # 103737-604
Supplier:  ACRO Biosystems
Supplier Number:  FO1C52H8100UG
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Cynomolgus monkey
  • Size:
    100 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    G7PR14-1
  • Carrier-Free:
    Y
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    FOLR-1|FOLR|FBP
  • Protein/Peptide Name:
    FOLR1
  • Purity:
    >90%
  • Molecular Weight:
    26.4 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping Temperature:
    Powder,RT
  • Cat. No.:
    103737-604

Specifications

About this item