Cynomolgus Recombinant Fc gamma RIII / CD16 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Rituximab at 5 μg/mL (100 μL/well) can bind Biotinylated Cynomolgus CD16, His,Avitag (BLI verified) (Cat. No. FC6-C82E0) with a linear range of 0.039-1.25 μg/mL (QC tested).; Captured Biotinylated Cynomolgus CD16, His,Avitag (BLI verified) (Cat. No. FC6-C82E0) on Biotin CAP - Series S sensor Chip can bind MabThera® (Rituximab) with an affinity constant of 0.63 μM as determined in a SPR assay (Biacore T200) (Routinely tested).; Loaded Biotinylated Cynomolgus CD16, His,Avitag (BLI verified) (Cat. No. FC6-C82E0) on SA Biosensor, can bind Rituximab with an affinity constant of 0.401 μM as determined in BLI assay (ForteBio Octet Red96e) (QC tested).
- Gene ID:Q8SPW2-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:FCGR3
- Protein/Peptide Name:Fc gamma RIII / CD16
- Purity:>95%
- Molecular Weight:25.7 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together