Bovine Recombinant FcRn (FCGRT and B2M) (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Bovine
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Biotinylated Bovine FCGRT&B2M Heterodimer Protein, His,Avitag (SPR verified) (Cat. No. FCN-B82W3) captured on Biotin CAP-Series S Sensor Chip can bind Herceptin® with an affinity constant of 2.59 nM as determined in a SPR assay (Biacore T200) (QC tested).
- Gene ID:Q3T119-1 (FCGRT) and P01888-1 (B2M)
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:FCGRT and B2M|FcRn
- Protein/Peptide Name:FcRn (FCGRT and B2M)
- Purity:>90%
- Molecular Weight:34.5 kDa (FCGRT) and 11.6 kDa (B2M)
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together