Recombinant Streptavidin, APC (Allophycocyanin), Thermo Scientific
PI21629
:
- Conjugation:APC (Allophycocyanin)
- Protein/Peptide Type:Recombinant
- Size:0.5 ml
- Protein/Peptide Name:Streptavidin
- Formulation:Lyophilized powder from approx. 0.5mg/mL protein in buffered saline solution containing bovine serum albumin
- Tested Applications:Fluorescence-Activated Cell Sorting and Histochemical Staining
- Cat. No.:PI21629
- Supplier no.:21629
Use to develop in-house non-radioactive assay systems.
Avidin can be used in ELISAs, immunohistochemical staining, Western blotting, hybridization, affinity chromatography, and fluorescent-activated cell sorting. The streptavidin conjugates are recommended for use where high binding affinity and specificity is required. NeutrAvidin® biotin-binding protein is a more economical alternative to both avidin and streptavidin. It features low nonspecific binding properties with no glycosylation. It also offers a neutral pI and lysine residues that remain available for derivatization or conjugation.