About this item
Corning 3D Clear Antibody Buffer used with immunolabeling in Corning 3D tissue labeling approach.
- Rapid tissue clearing
- Easy to Use
- No Special Equipment Required
- Compatible with IF, FP and other Fluorescent lables
- Microplate and Automation Compatible
- Reversible to Follow up with 2D H&E/IHC
The Corning 3D Clear Antibody Buffer is used with immunolabeling in the Corning 3D tissue labeling approach. This solution is comprised of PBS/0.2% Tween 20/Heparin/3% donkey serum/5% DMSO
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Related Information
Buffer Solutions Recipes for Immunolabeling
Additional Product Details
Corning 3D Clear Tissue Clearing Reagent