Order Entry
United States
ContactUsLinkComponent
 
VWR® Cleanroom Facemask, Class 100
  76415-182
undefined
VWR® Cleanroom Facemask, Class 100
  76415-182

 

  • Sterility:
    Non sterile
  • Color:
    White
  • Description:
    Class 100 facemask
  • Cat. No.:
    76415-182

 

 

Ear loop style facemasks are designed for use in Class 100 or higher cleanroom environments.

  • Low particles
  • High bacterial filtration efficiency (BFE)
  • Low breathable resistance
  • Packaged in a cleanroom environment
  • Non sterile

Masks are configured with quality non-woven Polypropylene/Polyethylene (ES) /Melt-blown Polypropylene (MBPP) materials.

 : Masks are packaged 100 per double bag, 20 bags per case. All bags are solvent safe.