Order Entry
United States
ContactUsLinkComponent
 
Volatile Organics Combination Analyte Mix for US EPA Method 624
  BDH0755-1ML
 :  
undefined
Volatile Organics Combination Analyte Mix for US EPA Method 624
  BDH0755-1ML
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Concentration:
    2000 µg/ml (2000 ppm)
  • Matrix:
    Methanol (P&T)
  • Pack type:
    Ampoule amber
  • Description:
    Volatile Organics Combination Analyte Mix for US EPA Method 624
  • Size:
    1 ml
  • Environmentally Preferable:
  • Cat. No.:
    BDH0755-1ML

 

 

Volatiles standard.

US EPA Method 624 is a general analytical method for the monitoring of VOCs in municipal and industrial wastewater by GC/MS.