Order Entry
United States
ContactUsLinkComponent
Polynuclear Aromatic Hydrocarbon (PAH) Mix for US EPA Methods 625
Polynuclear Aromatic Hydrocarbon (PAH) Mix for US EPA Methods 625
Catalog # BDH0736-1ML
Supplier:  VWR International
CAS Number:  
undefined
Polynuclear Aromatic Hydrocarbon (PAH) Mix for US EPA Methods 625
Catalog # BDH0736-1ML
Supplier:  VWR International
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Concentration:
    2000 µg/ml (2000 ppm)
  • Matrix:
    Methylene chloride:Benzene
  • Pack type:
    Ampoule amber
  • Description:
    Polynuclear Aromatic Hydrocarbon (PAH) Mix
  • Size:
    1 ml
  • Cat. No.:
    BDH0736-1ML

Specifications

About this item

Semivolatiles standard.

US EPA Method 625 is an analytical method for the monitoring of acid and base neutral extractable fractions in municipal and industrial wastewater by GC/MS.
US EPA Method 8100 is an analytical method for the monitoring of PNA hydrocarbons in aqueous and solid samples by GC/FID.
US EPA Method 8270 is a general analytical method for the monitoring of semivolatiles in environmental and waste samples by GC/MS.
CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.