Distek, Inc. has revolutionized benchtop scale bioreactors with the release of our BIOne 1250 bioprocess control station with BIOne single-use bioreactor for mammalian models. The sleek appearance and intuitive touchscreen interface are only the beginning. The BIOne 1250 is designed to be the easiest to use and most capable stand-alone controller on the market.
- Touchscreen interface
- Tmart media filling
- Email and text alerts
- Trend up to eight parameters
- Variable speed pumps
- Two and five litre working volumes
- Configurable for your lab
Control all bioreactor parameters directly from the BIOne 1250 interface, or, for remote control, use our optional OPC server and communicate with your choice of SCADA system.
From the pop-up tab of your selected parameter, control setpoints, calibrations, alarms, and more. Reduce development timelines by utilizing the live, dual graph trend screen. Analyze and compare current and previous runs. Track all optimized changes and export data to further analyze.